Merck
  • Phentermine and topiramate extended release (Qsymia™): first global approval.
Phentermine and topiramate extended release (Qsymia™): first global approval.
Drugs (2012-10-09)
Fiona Cameron, Glenn Whiteside, Kate McKeage
ABSTRACT
Vivus' proprietary oral capsule containing phentermine and extended-release (ER) topiramate has been approved in the US for the treatment of obesity. Phentermine is an appetite suppressant, while topiramate is an anti-epileptic medication. The once-daily formulation, known as Qsymia™, is designed to produce weight loss by decreasing appetite and increasing satiety. The product is also in clinical development for sleep apnoea syndrome and type 2 diabetes mellitus. This article summarizes the milestones in the development of phentermine/topiramate ER leading to this first approval for obesity.
Product Description
Supelco
Phentermine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®