Merck
  • Specificity of maltase to maltose in three different directions of reaction: hydrolytic, vanillyl alcohol glucoside and vanillyl alcohol isomaltoside synthesis.
Specificity of maltase to maltose in three different directions of reaction: hydrolytic, vanillyl alcohol glucoside and vanillyl alcohol isomaltoside synthesis.
Biotechnology progress (2012-08-29)
Aleksandra Dimitrijević, Dušan Veličković, Nenad Milosavić, Dejan Bezbradica
ABSTRACT
Vanillyl alcohol glucoside is very attractive molecule due to its very powerful physiological activity. In this article, a detailed kinetic study of transglucosylation of vanillyl alcohol was performed. It was demonstrated that this reaction is very efficient (selectivity factor is 149) and occurred by a ping-pong mechanism with inhibition by glucose acceptor. At low concentration of vanillyl alcohol one additional transglucosylation product was detected. Its structure was determined to be α-isomaltoside of vanillyl alcohol, indicating that vanillyl alcohol glucoside is a product of the first transglucosylation reaction and a substrate for second, so the whole reaction mechanism was proposed. It was demonstrated that the rate of isomaltoside synthesis is two orders of magnitude smaller than glucoside synthesis, and that maltase has interestingly high K(m) value to maltose when vanillyl alcohol glucoside is second transglucosylation substrate.
Sigma-Aldrich
Vanillyl alcohol, ≥98%, FG
Sigma-Aldrich
4-Hydroxy-3-methoxybenzyl alcohol, 98%