Merck
  • Prevalences of illicit drug use in people aged 50 years and over from two surveys.
Prevalences of illicit drug use in people aged 50 years and over from two surveys.
Age and ageing (2012-04-12)
Viola Fahmy, Stephani L Hatch, Matthew Hotopf, Robert Stewart
ABSTRACT
little is known about illicit drug use in older people. Prevalences may rise as populations for whom illicit drug use has been more common and acceptable become older. to quantify illicit drug use in people aged 50 years and over in England and inner London and to compare this between 50 and 64 and 65+ age groups. primary analyses used data from the 2007 Adult Psychiatric Morbidity Survey (APMS) and the 2008-10 South East London Community Health (SELCoH) Survey. Secondary analyses included additional data on 50-64 year olds from the 1993, 2000 and 2007 APMS, and on 65-74 year olds from the 2000 and 2007 APMS. cannabis was the drug most commonly used in all samples. Prevalences of use within the last 12 months in 50-64 and 65+ age groups were 1.8 and 0.4%, respectively, in England and 9.0 and 1.1%, respectively, in inner London. Prevalences of use at any time previously were 11.4, 1.7, 42.8 and 9.4%, respectively. Lifetime cannabis, amphetamine, cocaine and LSD use in 50-64 year olds had increased approximately tenfold in England from 1993. Lifetime and 12-month trends in tranquilisers were relatively stable. use of some illicit drugs, particularly cannabis, has increased rapidly in mid- and late-life.
Product Description
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®