Merck
  • Management of opioid-induced constipation in cancer patients: focus on methylnaltrexone.
Management of opioid-induced constipation in cancer patients: focus on methylnaltrexone.
Clinical drug investigation (2012-03-15)
Antonio Gatti, Alessandro Fabrizio Sabato
ABSTRACT
Opioids are one of the most widely used therapies for the palliative treatment of cancer pain; however, despite their proven analgesic efficacy, they are associated with several adverse effects. Associated with psychological distress and multiple concomitant clinical concerns, constipation is the most commonly occurring adverse effect of chronic opioid therapy in cancer patients. Whilst prophylaxis remains the first-line management option, methylnaltrexone is a recommended treatment option for opioid-related constipation if administration of laxatives is ineffective. Due to its inability to cross the blood-brain barrier, methylnaltrexone exerts a peripheral inhibition of opioid-related effects without influencing the opioid-induced central effects; as a result, the analgesic effect of opioids is unaffected. Moreover, multiple clinical trials, albeit not always conducted specifically in cancer patients, have demonstrated that up to 4 months' treatment with either intravenous or subcutaneous methylnaltrexone provides effective relief from opioid-related constipation and is well tolerated. Preliminary evidence indicates that the addition of methylnaltrexone to standard care for opioid-related constipation may also be advantageous from a pharmacoeconomic perspective. In addition, preliminary data suggest that methylnaltrexone could be associated with some further clinical benefits other than the treatment of opioid-related constipation, such as the improvement of gastric emptying, the relief of nausea/vomiting, and the reduction of the risk of regurgitation and pulmonary aspiration. This narrative review examines the most recent evidence and evaluates the current role of methylnaltrexone in the management of opioid-related constipation, and its potential efficacy in cancer patients. The pharmacokinetics, pharmacodynamics, efficacy and tolerability of methylnaltrexone are discussed.
Sigma-Aldrich
Methylnaltrexone bromide, ≥97% (HPLC)