Merck
  • Signs and symptoms of ocular surface status in glaucoma patients switched from timolol 0.5% to brinzolamide 1%/timolol 0.5% fixed combination: a 6-month efficacy and tolerability, multicenter, open-label prospective study.
Signs and symptoms of ocular surface status in glaucoma patients switched from timolol 0.5% to brinzolamide 1%/timolol 0.5% fixed combination: a 6-month efficacy and tolerability, multicenter, open-label prospective study.
Expert opinion on pharmacotherapy (2011-03-10)
Gemma Caterina Maria Rossi, Carmine Tinelli, Gian Maria Pasinetti, Marina Fusetti, Claudio Pallavicini, Massimo Stringa, Sauro Vacchi, Francesco Stringa, Paolo Emilio Bianchi
ABSTRACT
To examine the impact of switching glaucoma patients from timolol 0.5% to brinzolamide 1%/timolol 0.5% fixed combination (Brinz/Tim FC) on quality of life and on ocular surface status; to assess efficacy after the switch. 6-month, multicenter, open-label, prospective, switch study in 119 early to moderate glaucoma patients. Intraocular pressure (IOP), tear film break-up time (TF-BUT), fluorescein staining and Glaucoma Symptom Scale (GSS) questionnaire were recorded at baseline and after 6 months. Median (interquartile range) IOP significantly decreased from 20 to 16 mmHg, independent of sex and age. Most patients (95.8%) reached an IOP < 18 mmHg. TF-BUT improved, with a negative weak correlation to age at baseline and at 6 months. The percentage of patients with no fluorescein staining improved. The quality of life recorded by GSS also improved, being related both to age and corneal staining. Brinz/Tim FC is effective and well tolerated. In this study, patients switched to Brinz/Tim FC obtained further reduction in IOP with no effects on ocular surface status and improved quality of life perception: a better quality of life could determine a better adherence to prescribed therapy.
Sigma-Aldrich
Brinzolamide, ≥98% (HPLC)