Merck
  • Palladium(II) complex with S-allyl-L-cysteine: new solid-state NMR spectroscopic measurements, molecular modeling and antibacterial assays.
Palladium(II) complex with S-allyl-L-cysteine: new solid-state NMR spectroscopic measurements, molecular modeling and antibacterial assays.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-11-06)
Marcelle B M Spera, Frederico A Quintão, Diego K D Ferraresi, Wilton R Lustri, Alviclér Magalhães, André L B Formiga, Pedro P Corbi
ABSTRACT
Nuclear magnetic resonance studies, molecular modeling and antibacterial assays of the palladium(II) complex with S-allyl-L-cysteine (deoxyalliin) are presented. Studies based on solid and solution 13C and 15N nuclear magnetic resonance (NMR) spectroscopy confirmed that the palladium(II) complex preserved the same structural arrangement in both states, with no modifications on coordination sphere when dissolved in water. Density functional theory (DFT) studies stated that the trans isomer is the most stable one. Antibacterial activities of S-allyl-L-cysteine and its palladium(II) complex were evaluated by antibiogram assays using the disc diffusion method. The palladium(II) complex showed an effective antibacterial activity against Staphylococcus aureus (Gram-positive), Escherichia coli and Pseudomonas aeruginosa (Gram-negative) bacterial cells.
Sigma-Aldrich
S-Allyl-L-cysteine, ≥98% (HPLC)