Merck
  • Interaction of the hexamethonium derivative W84, an allosteric effector at muscarinic cholinoreceptors, with rat brain nicotinic binding sites.
Interaction of the hexamethonium derivative W84, an allosteric effector at muscarinic cholinoreceptors, with rat brain nicotinic binding sites.
Pharmacology & toxicology (1990-10-01)
F Leithäuser, K Mohr, C M Staschen
ABSTRACT
The hexamethonium derivative W84 (hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)-ammonium bromide]) combined with atropine has an overadditive protective action against organophosphorus intoxications. It affects allosterically the binding of (-) [3H]N-methylscopolamine [(3H]NMS) to muscarinic cholinoceptors. Because nicotinic receptors are involved in organophosphorus intoxications, the interaction of W84 with nicotinic cholinoceptors was investigated. (-) [3H]nicotine (2.5 nM) was used to label nicotinic binding sites in rat brain membranes in 50 nM Tris, pH 7.3 at 23 degrees. Under control conditions, (-) [3H]nicotine-binding revealed a KD of 4 X 10(-9) M and a Bmax of 53 fmol/mg membrane protein. W84 inhibited (- ) [3H]nicotine-binding with an IC50 of 3 X 10(-5) M by reducing the binding affinity. The IC50 of hexamethonium was 20 X 10(-5) M. At 10(-4) M, W84 did not affect the dissociation rate of (-)[3H]nicotine, suggesting a lack of allosteric activity. For sake of comparison, the action of W84 was checked on [3H]NMS-binding (control: KD approximately 1 X 10(-9) M, Bmax approximately 500 fmol/mg prot). W84 inhibited the binding of [3H]NMS (0.5 nM) with an IC50 of 1.5 X 10(-9) M. At 10(-4) M, W84 prevented [3H]NMS-dissociation almost completely, thus displaying the allosteric action at muscarinic cholinoceptors. In conclusion, the results of the (-)[3H]nicotine-binding experiments point to a pure competitive action of W84 at nicotine cholinoceptors, lacking any allosteric effect. This competitive action may contribute to the protective effect of W84 in organophosphorus poisoning.