Merck
  • Effects of a cognitive modulator in the theta and alpha asymmetry during a typewriting task: a sensorimotor integration perspective.
Effects of a cognitive modulator in the theta and alpha asymmetry during a typewriting task: a sensorimotor integration perspective.
Arquivos de neuro-psiquiatria (2009-06-24)
Marlo Cunha, Sergio Machado, Luiz Cláudio Miana, Dionis Machado, Victor Hugo Bastos, Bruna Velasques, Maurício Cagy, Luis F Basile, Roberto Piedade, Pedro Ribeiro
ABSTRACT
This study aimed to elucidate cortical mechanisms and to identify the areas where occur such mechanisms due to interaction between bromazepam and motor learning. The sample was composed of 45 healthy subjects randomly distributed in 3 groups: placebo (n=15), bromazepam 3 mg (n=15) or bromazepam 6 mg (n=15). To perform the experimental task, subjects sat comfortably at a distance of approximately 20 cm from the typewriter. The typewriter keyboard was covered with a wooden box to avoid visual information about the hands' position. The typewriting task was performed concomitantly with EEG recording. ANOVA two-way results indicated a decreased asymmetry in sensorimotor areas in the experimental groups. Our interpretation is that moderate doses of bromazepam may improve performance on tasks with predictable elements to promote stability of psychomotor functions, but may also impair performance on tasks executed in unpredictable environments.
Product Description
Supelco
Bromazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®