Merck
  • Effect of RGD-immobilized dual-pore poly(L-lactic acid) scaffolds on chondrocyte proliferation and extracellular matrix production.
Effect of RGD-immobilized dual-pore poly(L-lactic acid) scaffolds on chondrocyte proliferation and extracellular matrix production.
Artificial organs (2009-01-10)
Hyun Jung Jung, Kwideok Park, Jae-Jin Kim, Jin Ho Lee, Ki-Ok Han, Dong Keun Han
ABSTRACT
Interactions between cell and polymer surface have great implications in tissue engineering. In this study, chondrocyte proliferation and matrix production were examined using porous poly(L-lactide) (PLLA) scaffolds that have different surface characteristics. PLLA scaffolds were prepared using a gas-foaming method, and subjected to surface modifications through plasma treatment and subsequent in situ grafting of hydrophilic acrylic acid (AA). To immobilize peptide ligands, the AA-grafted PLLA scaffolds (PLLA-PAA) were further reacted with either Gly-Arg-Asp-Gly (GRDG) or Gly-Arg-Gly-Asp (GRGD) to produce PLLA-PAA-GRDG or PLLA-PAA-GRGD scaffold, respectively. The average porosities of the scaffolds were more than 90%, and their pore sizes ranged from 200 approximately 300 to 10 approximately 50 microm for large and small pores, respectively. The concentrations of each bound component were 2.14 x 10(-4) mmol/cm(2) for AA, 1.87 nmol/g for GRDG, and 1.68 nmol/g for GRGD. When chondrocytes were seeded onto the different PLLA scaffolds, cell adhesion and proliferation were highly affected as the substrate types vary. The RGD-immobilized scaffolds resulted in higher cellularity and better accumulation of total glycosaminoglycan than the others. Histological staining of Safranin O showed that the deposited extracellular matrix was more intense and widely distributed in the PLLA-PAA-GRGD scaffold. The present data suggest that immobilization of RGD peptide on the AA-grafted PLLA scaffold can be an effective tool for chondrocyte attachment and proliferation, and that it may also be helpful to facilitate cartilaginous tissue formation.