Merck
  • Metabolism of echinacoside, a good antioxidant, in rats: isolation and identification of its biliary metabolites.
Metabolism of echinacoside, a good antioxidant, in rats: isolation and identification of its biliary metabolites.
Drug metabolism and disposition: the biological fate of chemicals (2008-11-22)
Cunqin Jia, Haiming Shi, Wei Jin, Ke Zhang, Yong Jiang, Mingbo Zhao, Pengfei Tu
ABSTRACT
Echinacoside (ECH) is one of the major active phenylethanoid glycosides (PEGs) in famous traditional Chinese medicine, Herba Cistanches. Although it has various bioactivities, such as antioxidation, neuroprotection, and hepatoprotection, knowledge about its metabolic fate is scant. In the present study, eight phase II metabolites, 3,4 -O-dimethyl-ECH-3 -O-beta-d-glucuronide (M1); 4,4 -O-dimethyl-ECH-3 -O-beta-d-glucuronide (M2); 3,4 -O-dimethyl-ECH-4-O-sulfate ester (M3); 4,4 -O-dimethyl-ECH-3-O-sulfate ester (M4); 3,3 -O-dimethyl-ECH (M5); 3,4 -O-dimethyl-ECH (M6); 4,3 -O-dimethyl-ECH (M7); and 4,4 -O-dimethyl-ECH (M8), were isolated from rat bile sample after intravenous administration of ECH and identified by mass spectra and NMR spectroscopy, including (1)H NMR, (13)C NMR, nuclear Overhauser effect difference spectroscopy, and two-dimensional NMR (heteronuclear single quantum correlation, heteronuclear multiple-bond correlation spectroscopy, gradient-selected correlation spectroscopy, and nuclear Overhauser effect spectroscopy). Among them, M5 to M8 were O-di-methylated conjugates; M1 and M2 and M3 and M4 were O-dimethyl glucuronides and O-dimethyl sulfates, respectively. In the three types of metabolites of rat, the major metabolites were the methyl ethers and the glucuronides, whereas the sulfates were minor. The regioselectivity of conjugation for ECH and metabolic pathway of ECH were proposed, which gave insight into the mechanism of ECH for its bioactivities in vivo.
Sigma-Aldrich
Echinacoside, 98% (HPLC)
Supelco
Echinacoside, analytical standard