Merck
  • Interaction of plant cannabinoids with the multidrug transporter ABCC1 (MRP1).
Interaction of plant cannabinoids with the multidrug transporter ABCC1 (MRP1).
European journal of pharmacology (2008-07-16)
Michelle L Holland, John D Allen, Jonathon C Arnold
ABSTRACT
The ATP-binding cassette (ABC) transporter ABCC1, or multidrug resistance-related protein 1 (MRP1) is implicated in Phase II metabolism and multidrug resistance as it effluxes substrate anticancer drugs. As cannabinoids inhibit two related ABC transporters, P-glycoprotein and ABCG2, here we examined whether they also inhibit ABCC1. Indeed, the cannabinoids enhanced the intracellular accumulation of two ABCC1 substrates, Fluo3 and vincristine, in ovarian carcinoma cells over-expressing ABCC1 (2008/MRP1) with a rank order of potency: cannabidiol>cannabinol>Delta(9)-tetrahydrocannabinol. Cannabinoid inhibition of ABCC1 was confirmed using insect cell membrane MRP1 ATPase assays. These results demonstrate that cannabinoids inhibit ABCC1.
Product Description
Supelco
Cannabinol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®