Merck
  • Probing the active site of a diels-alderase ribozyme by photoaffinity cross-linking.
Probing the active site of a diels-alderase ribozyme by photoaffinity cross-linking.
Journal of the American Chemical Society (2008-06-12)
Richard Wombacher, Andres Jäschke
ABSTRACT
The active site of a Diels-Alderase ribozyme is located in solution by photoaffinity cross-linking using a productlike azidobenzyl probe. Two key nucleotides are identified that contact the Diels-Alder product in a conformation-dependent fashion. The design of such probes does not require knowledge of the three-dimensional structure of the ribozyme, and the technique yields both static and dynamic structural information. This work establishes photoaffinity cross-linking as an empirical approach that is applied here for the first time to an artificial ribozyme.
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in tert-butyl methyl ether
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in 2-methyltetrahydrofuran, ≥95.0% (HPLC)