Merck
Substrate flexibility of a 2,6-dideoxyglycosyltransferase.
Chemical communications (Cambridge, England) (2006-10-19)
David L Jakeman, Charles N Borissow, Cathy L Graham, Shannon C Timmons, Taryn R Reid, Ray T Syvitski
ABSTRACT
We report the first 2,6-dideoxysugar-O-glycosyltransferase with substrate flexibility at the 2 position, confirm the function of a putative NDP-hexose 2,3-dehydratase in the jadomycin B biosynthetic gene cluster and deduce the substrate flexibility of downstream enzymes in l-digitoxose assembly, enabling reprogramming of biosynthetic gene clusters to modify sugar substituents.