Merck
  • Synthesis of 2,4-disubstituted 5-aminoimidazoles using microwave irradiation.
Synthesis of 2,4-disubstituted 5-aminoimidazoles using microwave irradiation.
Journal of combinatorial chemistry (2006-07-11)
Chai Hoon Soh, Wai Keung Chui, Yulin Lam
ABSTRACT
A microwave-assisted parallel synthesis of 2,4-disubstituted 5-aminoimidazoles has been developed. Significant rate enhancement was observed for all steps in the three-step protocol. The overall reaction time was shortened to 25 min, as compared to 53 h for the conventional procedures. A representative set of 2,4-disubstituted 5-aminoimidazoles was prepared using commercially available parallel reactors.
Sigma-Aldrich
5-Aminoindazole, 97%