Merck
  • Enrichment and analysis of peptide subsets using fluorous affinity tags and mass spectrometry.
Enrichment and analysis of peptide subsets using fluorous affinity tags and mass spectrometry.
Nature biotechnology (2005-03-16)
Scott M Brittain, Scott B Ficarro, Ansgar Brock, Eric C Peters
ABSTRACT
Although mass spectrometry has become a powerful tool for the functional analysis of biological systems, complete proteome characterization cannot yet be achieved. Instead, the sheer complexity of living organisms demands fractionation of cellular extracts to enable more targeted analyses. Here, we introduce the concept of "fluorous proteomics," whereby specific peptide subsets from samples of biological origin are tagged with perfluorinated moieties and subsequently enriched by solid-phase extraction over a fluorous-functionalized stationary phase. This approach is extremely selective, yet can readily be tailored to enrich different subsets of peptides. Additionally, this methodology overcomes many of the limitations of traditional bioaffinity-based enrichment strategies, while enabling new affinity enrichment schemes impossible to implement with bioaffinity reagents. The potential of this methodology is demonstrated by the facile enrichment of peptides bearing particular side-chain functionalities or post-translational modifications from tryptic digests of individual proteins as well as whole cell lysates.
Sigma-Aldrich
O-Methylisourea hemisulfate salt, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
o-Methylisourea bisulfate, 99%