Merck
G-protein-coupled receptor allosterism: the promise and the problem(s).
Biochemical Society transactions (2004-10-21)
A Christopoulos, L T May, V A Avlani, P M Sexton
ABSTRACT
Allosteric modulators of G-protein-coupled receptors interact with binding sites that are topographically distinct from the orthosteric site recognized by the receptor's endogenous agonist. Allosteric ligands offer a number of advantages over orthosteric drugs, including the potential for greater receptor subtype selectivity and a more 'physiological' regulation of receptor activity. However, the manifestations of allosterism at G-protein-coupled receptors are quite varied, and significant challenges remain for the optimization of screening methods to ensure the routine detection and validation of allosteric ligands.
Product Description
Alcuronium chloride, European Pharmacopoeia (EP) Reference Standard