Merck
Degradation of N-acetyl tryptophan by low-energy (<12 eV) electrons.
Journal of the American Chemical Society (2004-09-24)
Hassan Abdoul-Carime, Sascha Gohlke, Eugen Illenberger
ABSTRACT
Secondary low-energy electrons are abundantly created during the early moments following the deposition of energy by radiation into cells. Here we show the ability of slow (<12 eV) electrons to effectively decompose gas-phase N-acetyl tryptophan (NAT) which can model a simple protein. The fragmentation of NAT, initiated via a resonant electron-molecule interaction exclusively at the peptide bridge, produces a large variety of negative species. The present findings contribute to the molecular description of the initial step in the radiation-induced damage.
Sigma-Aldrich
N-Acetyl-L-tryptophan
Supelco
N-Acetyl-L-tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material