Merck
  • Interaction between ketopantolactone and chirally modified Pt investigated by attenuated total reflection IR concentration modulation spectroscopy.
Interaction between ketopantolactone and chirally modified Pt investigated by attenuated total reflection IR concentration modulation spectroscopy.
Journal of the American Chemical Society (2003-10-30)
Norberto Bonalumi, Thomas Bürgi, Alfons Baiker
ABSTRACT
The combination of ATR-IR and modulation spectroscopy allowed for the study of the interaction of ketopantolactone with Pt/Al2O3 films chirally modified by cinchonidine under hydrogenation conditions. The spectra reveal a significant influence of ketopantolactone on the adsorption of the modifier and indicate a N-H-O hydrogen bond between modifier and reactant. The latter was corroborated by a comparative study with N-methyl cinchonidine chloride modified Pt/Al2O3.
Sigma-Aldrich
Dihydro-4,4-dimethyl-2,3-furandione, 97%