Merck
  • [Advances in modulation of mammalian sperm function by fertilization promoting peptide].
[Advances in modulation of mammalian sperm function by fertilization promoting peptide].
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics (2003-04-04)
Yongxin Ma, Sizhong Zhang
ABSTRACT
Both fertilization promoting peptide and adenosine stimulate capacitation but inhibit spontaneous acrosome loss by modulation of the adenylyl cyclase (AC)/cAMP signal transduction pathway. This is a review aimed at analyzing the function of fertilization promoting peptide during this process. The possible molecular basis is also discussed.