Merck
  • A chronic inhalation toxicity/oncogenicity study of methylethylketoxime in rats and mice.
A chronic inhalation toxicity/oncogenicity study of methylethylketoxime in rats and mice.
Inhalation toxicology (2001-11-07)
P E Newton, W L Wooding, H F Bolte, M J Derelanko, J F Hardisty, W E Rinehart
ABSTRACT
To evaluate the oncogenic potential of methylethylketoxime (MEKO), CD-1 mice (50/sex/group) and F-344 rats (50/sex/group) were coexposed 6 h/day, 5 days/wk for 18 mo (mice) or 26 mo (rats) via whole-body inhalation exposures to target vapor concentrations of 0, 15, 75, and 375 ppm (actual concentrations of 0, 15 +/- 1, 75 +/- 2, or 374 +/- 10 ppm). Satellite groups of rats and mice (10/sex/group/interval) were exposed for 12 mo (mice) and 3, 12, or 18 mo (rats) to evaluate chronic toxicity. Methyl ethyl ketone (MEK), a possible hydrolysis product of MEKO, was present at less than 1%. Treatment-related effects included increased body weight (male rats only), methemoglobin formation, hematology and clinical chemistry changes, increased liver weight, and increased spleen and testes weights (rats only). A high incidence of cataracts and corneal dystrophy occurred in both control and MEKO-exposed rats, with an earlier appearance and slightly higher incidence for these ocular lesions in MEKO-exposed animals compared to controls. Degenerative and reparative changes of the olfactory epithelium in the nasal turbinates, primarily limited to the dorsal meatus, occurred in both rats (75 and 374 ppm) and mice (15, 75, and 374 ppm). In addition, in the mice, liver changes included increased incidences of pigment in reticuloendothelial cells, centilobular hypertrophy, granulomatous inflammation, and a slightly increased incidence of necrosis (75 and 374 ppm). An increase in hepatocellular carcinomas occurred in male mice at 374 ppm. Additional MEKO-related findings in the rat included congestion of the spleen with pigment in reticuloendothelial cells and extramedullary hematopoiesis and a decreased incidence of lymphoreticular mononuclear cell leukemia. Effects observed in the liver of the rats included decreases in the incidence of both peribiliary fibrosis and hyperplasia/proliferation of the biliary duct, an increase of spongiosis hepatis in males, and an increase in the incidence of intracytoplasmic vacuoles and hepatocellular basophilic foci. The effects on the liver were generally most profound in the high-exposure groups and, with the exception of the spongiosis hepatis, occurred in both sexes. An increase in hepatocellular adenomas occurred in the male rats at 75 and 374 ppm, and hepatocellular carcinomas in the male rats at 374 ppm. In both species, the liver tumors appeared relatively late in the life of the animals, with no significant increase in tumors at 12 mo of exposure in mice and at 18 mo of exposure in rats. Lifespan shortening was not observed, as MEKO-exposed animals survived generally as well as, or slightly better than, the controls.
Sigma-Aldrich
2-Butanone oxime, 99%