Merck
  • Combination of N-methyl-1-deoxynojirimycin and ischemic preconditioning markedly reduces the size of myocardial infarcts in rabbits.
Combination of N-methyl-1-deoxynojirimycin and ischemic preconditioning markedly reduces the size of myocardial infarcts in rabbits.
Japanese circulation journal (2001-07-12)
D J Wu, S Minatoguchi, Y Uno, M Arai, N Wang, Y Nishida, K Hashimoto, Y Hashimoto, G Takemura, T Fujiwara, H Fujiwara
ABSTRACT
N-methyl-1-deoxynojirimycin (NMDN), an a-glucosidase inhibitor, reduces myocardial infarct size by reducing the glycogenolytic rate through inhibition of the alpha-1,6-glucosidase of glycogen-debranching enzyme in the heart, in addition to possessing an antihyperglycemic action by blocking alpha-1,4-glucosidase in the intestine. Ischemic preconditioning (PC), which markedly reduces the size of the myocardial infarct, is known to reduce the activity of phosphorylase and reduce the glycogenolytic rate. Therefore, it was hypothesized that a combination of pharmacological inhibition of glycogenolysis by an alpha-1,6-glucosidase inhibitor, NMDN, and PC could markedly reduce myocardial infarct size more than NMDN or PC alone. Japanese white rabbits without collateral circulation were subjected to a 30-min coronary occlusion followed by 48-h reperfusion. The infarct sizes as a percentage of area at risk were significantly reduced by pre-ischemic treatment with either 100mg/kg of NMDN or PC of 5 min ischemia and 5 min reperfusion alone (15.9+/-2.0%, n=8, and 10.3+/-1.2%, n=8, respectively) as compared with the control (43.9+/-2.2%, n=8). However, the combination of 100mg/kg of NMDN and PC significantly reduced the infarct size (4.9+/-1.2, n=8) compared with NMDN or PC alone. Another 40 rabbits, also given 100mg of NMDN, PC, NMDN+PC or saline before ischemia (n=10 in each group), were killed for biochemical analysis after 30 min of ischemia. NMDN and PC preserved the glycogen content and attenuated the lactate accumulation, respectively, as compared with the control. However, the combination of NMDN and PC preserved significantly more glycogen and significantly reduced lactate accumulation than either NMDN or PC alone. The combination of NMDN and PC markedly reduced the myocardial infarct size more than either process alone. The marked preservation of glycogen and marked attenuation of lactate accumulation by the combination of NMDN and PC suggest that the mechanism for this effect of NMDN+PC is related to the inhibition of glycogenolysis.
Sigma-Aldrich
N-Methyl-1-deoxynojirimycin, ≥98%