Merck
  • Iridium-catalyzed C-C coupling via transfer hydrogenation: carbonyl addition from the alcohol or aldehyde oxidation level employing 1,3-cyclohexadiene.
Iridium-catalyzed C-C coupling via transfer hydrogenation: carbonyl addition from the alcohol or aldehyde oxidation level employing 1,3-cyclohexadiene.
Organic letters (2008-02-08)
John F Bower, Ryan L Patman, Michael J Krische
ABSTRACT
Under hydrogen autotransfer conditions employing a catalyst derived from [Ir(cod)Cl]2 and BIPHEP, 1,3-cyclohexadiene (CHD) couples to benzylic alcohols 1a-9a to furnish carbonyl addition products 1c-9c, which appear as single diastereomers with variable quantities of regioisomeric adducts 1d-9d. Under related transfer hydrogenation conditions employing isopropanol as terminal reductant, identical carbonyl adducts 1c-9c are obtained from the aldehyde oxidation level. Isotopic labeling studies corroborate a mechanism involving hydrogen donation from the reactant alcohol or sacrificial alcohol (i-PrOH).