MilliporeSigma
  • Determination on functional basis of presynaptic alpha 2-adrenoceptor subtypes in guinea-pig duodenum.
Determination on functional basis of presynaptic alpha 2-adrenoceptor subtypes in guinea-pig duodenum.
Neuroscience letters (1996-05-24)
R Colucci, C Blandizzi, D Carignani, G Lazzeri, G Natale, F Crema, M Del Tacca
ABSTRACT
The effects of several alpha 2-adrenoceptor agonists and antagonists were examined on the cholinergic twitch contractions evoked by electrical field stimulation of guinea-pig duodenum. Oxymetazoline, xylazine, noradrenaline, alpha-methyl-noradrenaline or medetomidine (0.01-30 microM) were nearly equieffective in inhibiting duodenal twitch responses. The effects of xylazine were competitively counteracted by antagonists tested (0.03-10 microM) with the following order of potency: RX 821002 = idazoxan > rauwolscine = yohimbine = BRL 44408 > prazosin = ARC 239 = BRL 41992. According to the current classification, it is suggested that alpha 2-heteroadrenoceptors involved in the modulation of duodenal cholinergic neurotransmission belong to the alpha ZD subtype.
Sigma-Aldrich
ARC 239 dihydrochloride hydrate, ≥98% (HPLC)