Merck
  • Effects of charge, volume, and surface on binding of inhibitor and substrate moieties to acetylcholinesterase.
Effects of charge, volume, and surface on binding of inhibitor and substrate moieties to acetylcholinesterase.
Journal of medicinal chemistry (1985-09-01)
S G Cohen, S B Chishti, J L Elkind, H Reese, J B Cohen
ABSTRACT
Reversible inhibitors for acetylcholinesterase, AcChE, have been studied. Sterically similar alcohols with tetra-substituted uncharged beta groups, (CH3)3SiCH2CH2OH (I), (CH3)3CCH2CH2OH (IA), and CH3S(O2)CH2CH2OH (VII), bind similarly, KI = 3-9 mM, and each binds similarly to its acetate substrate; cationic analogues, (CH3)3N+CH2CH2OH (IB) and (CH3)2S+CH2CH2OH (II), bind similarly to each other, KI = 0.4 mM, similar to Km values of their acetate substrates, and more strongly than the uncharged alcohols by approximately 1.5 kcal/mol. In comparisons of VII with CH3SO2CH3, II with (CH3)3S+, and IB with (CH3)4N+, hydroxyethyl leads to more favorable binding than methyl by approximately 0.8 kcal/mol, despite lower hydrophobicity. Two hydrophobic methyl groups, in comparison of IA with butanol, and two hydrophilic sulfone O atoms, in comparison of VII with 2-(methylthio)ethanol, increase binding similarly, by 1.0 kcal/mol. Conversion of (CH3)3S+ to (CH3)3S+O also improves binding. However, (CH3)3N+O- does not bind to AcChE, and conversion of 1-(dimethylammonio)-4-pentanone and 2-(dimethylammonio)ethyl acetate to their N-oxides, changes of identical to N+H to identical to N+--O-, decreases binding by 1.5 kcal/mol. Although the -COCH3 group in esters with well-binding beta substituents makes essentially no contribution to binding over that of the alcohols, in esters with weakly bound beta substituents, (CH3)2N+(O-), CH3N+H2, CH3S(O), CH3CH2, and CH3S binding is dominated by the ester -COCH3 group, with values of Km approximately 16 mM.
Sigma-Aldrich
Trimethylsulfonium iodide, 98%
Sigma-Aldrich
2-(Trimethylsilyl)ethanol, 96%