Merck
  • Xanthine oxidase-catalyzed DNA binding of dihydrodiol derivatives of nitro-polycyclic aromatic hydrocarbons.
Xanthine oxidase-catalyzed DNA binding of dihydrodiol derivatives of nitro-polycyclic aromatic hydrocarbons.
Biochemical and biophysical research communications (1986-11-26)
K K Colvert, P P Fu
ABSTRACT
Xanthine oxidase, a mammalian nitroreductase, catalyzed the covalent binding of a series of nitro-polycyclic aromatic hydrocarbons (nitro-PAHs) trans-dihydrodiols to DNA. Some of the trans-dihydrodiols bound to DNA to a greater extent than their parent nitro-PAHs; however, when the dihydrodiol moiety was peri to the nitro substituent low levels of binding were observed. These data illustrate that ring-oxidation and hydrolysis of nitro-PAHs to their trans-dihydrodiols followed by nitroreduction is a potential metabolic pathway leading to DNA adducts in mammals.
Sigma-Aldrich
9-Nitroanthracene, 93%
9-Nitroanthracene, BCR®, certified reference material