Merck
  • A method for screening penicillin G acylase-producing bacteria by means of 2-nitro-5-phenylacetaminobenzoic acid test paper.
A method for screening penicillin G acylase-producing bacteria by means of 2-nitro-5-phenylacetaminobenzoic acid test paper.
Analytical biochemistry (1986-08-01)
Q J Zhang, L F Zhang, H X Han, Y Y Zhang
ABSTRACT
A simple, rapid assay for screening penicillin G acylase-producing bacteria is presented. The method is based on the formation of yellow 2-nitro-5-aminobenzoic acid by penicillin G acylase acting on 2-nitro-5-phenylacetaminobenzoic acid (NIPAB). NIPAB test paper is briefly applied to bacterial colonies on the agar surface, which are subsequently scored individually on the paper by color; bright yellow indicates the presence of penicillin G acylase, natural color its absence. The present method is suitable not only for screening penicillin G acylase-production by a variety of bacteria but also for detection from a large number of transformant colonies of clones containing a gene encoding for the enzyme.
Sigma-Aldrich
6-Nitro-3-(phenylacetamido)benzoic acid, ≥98%