Merck
  • Nanorod vanadatesulfuric acid as a novel, recyclable and heterogeneous catalyst for the one-pot synthesis of tetrahydrobenzopyrans.
Nanorod vanadatesulfuric acid as a novel, recyclable and heterogeneous catalyst for the one-pot synthesis of tetrahydrobenzopyrans.
Journal of nanoscience and nanotechnology (2013-08-02)
Masoud Nasr-Esfahani, Tooba Abdizadeh
ABSTRACT
Vanadatesulfuric acid (VSA), as a novel and heterogeneous nanorod catalyst, was used for an efficient synthesis of tetrahydrobenzo[b]pyrans using an aldehydes, 1,3-cyclohexanediones or beta-ketoester and malononitrile in C2H5OH/H2O mixture under reflux conditions. VSA is prepared via the reaction of sodium metavanadate and chlorosulfonic acid in high purity. The catalyst was characterized by FT-IR, XRD, TEM and EDAX analysis. Compared to the conventional method, this method consistently has the advantage of high yields, simple workup, short reaction times and reusability of the catalyst.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC