Merck
  • In vitro ALP and osteocalcin gene expression analysis and in vivo biocompatibility of N-methylene phosphonic chitosan nanofibers for bone regeneration.
In vitro ALP and osteocalcin gene expression analysis and in vivo biocompatibility of N-methylene phosphonic chitosan nanofibers for bone regeneration.
Journal of biomedical nanotechnology (2013-06-28)
Pallab Datta, Paulomi Ghosh, Kuntal Ghosh, Pritiprasanna Maity, Sintu Kumar Samanta, Sudip Kumar Ghosh, Pradeep Kumar Das Mohapatra, Jyotirmoy Chatterjee, Santanu Dhara
ABSTRACT
Most polymeric nanofibers used for bone tissue engineering lack adequate functional groups for bioactivity. This study explores the potential of nanofibers of phosphate functionalized derivative of chitosan-N-methylene phosphonic chitosan (NMPC) for bone tissue engineering. Nanofibers were fabricated by electrospinning of NMPC/PVA blend solutions. NMPC/PVA nanofibers exhibited 172% higher viability of MG-63 cells compared to pure PVA nanofibers. ALP and Collagen type I genes revealed higher expression in NMPC nanofibers on day 3 whereas osteocalcin gene was expressed on day 7. In rabbit tibial defects, NMPC based electrospun graft showed presence of no adverse tissue reaction by histological examination while radiological examination suggested acceleration of bone healing by 300% compared to defects without any scaffold. Thus it is concluded NMPC based nanofibers may have potential for bone grafting applications.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous