Merck
[Chemical constituents of Rhododendron seniavinii].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-15)
Qing-Qing Wang, Ying Zhang, Wen-Cai Ye, Guang-Xiong Zhou
ABSTRACT
To study the chemical constituents of Rhododendron seniavinii. Compounds were isolated from the aqueous extract of the leaves of R. seniavinii by using Sephadex LH-20, ODS open column chromatography and other means. Their structures were elucidated according to spectral data and physiochemical properties. Thirteen compounds were isolated from R. seniavinii and identified as 5-methoxydehydroconiferyl alcohol (1), dehydroconiferyl alcohol (2), (-)-syringaresinol (3), (-)-lyoniresinol (4), (+)-lyoniresinol 3alpha-O-beta-D-glucopyranoside (5), (-)-lyoniresinol 3alpha-O-beta-D-glucopyranoside (6), 3,4,5-trimethoxyphenyl-1-O-beta-D-glucopyranoside (7), nikoenoside (8), 3,5,7-trihydroxychromone-3-0-alpha-L-rhamnopyranoside (9), 3,4,5-trimethoxyphenol (10), scopoletin (11), scopolin (12) and quercitrin (13). Compounds 1-12 were obtained from this plant for the first time.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Sephadex® G-100, BioReagent, for molecular biology, DNA grade, medium
Supelco
Dextran, analytical standard, for GPC, 150,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Sephadex® G-75, Medium
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-25, Fine
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Sephadex® G-25, BioReagent, for molecular biology, DNA grade, Superfine
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000