Merck
  • Enhanced fluorescence, morphological and thermal properties of CdSe/ZnS quantum dots incorporated in silicone resin.
Enhanced fluorescence, morphological and thermal properties of CdSe/ZnS quantum dots incorporated in silicone resin.
Journal of nanoscience and nanotechnology (2013-05-08)
Nguyen Ngoc Trung, Quynh-Phuong Luu, Bui Thanh Son, Le Hoang Sinh, Jin-Young Bae
ABSTRACT
Our research focused on the morphological and optical properties of core/shell cadmium selenide/zinc sulfide (CdSe/ZnS) quantum dots incorporated in silicone resin. After dispersing ligand-coated quantum dots into Dow Corning two-component silicone resins (OE6630A and OE6630B at 1:4 mixing ratio by weight), the resins were cured at 150 degrees C for 1.5 hours to produce the quantum dot-silicone resin nanocomposites. The optical, morphological and thermal properties of the quantum dot incorporated in silicone resin were investigated by ultraviolet-visible, fluorescence, atomic force microscopy, field emission scanning electron microscopy, differential scanning calorimetry and thermogravimetric analysis. When the quantum dots, originally coated with trioctylamine ligand, were transferred from a chloroform solvent to methyl phenyl silicone oil and silicone resins of high viscosity, the quantum dots showed increased turbidity and lowered fluorescence intensity. Fluorescence enhancement was investigated by using various functional ligands such as poly(1, 1-dimethyl silazane) (multi-silazane), hexamethylenediamine (diamine), cysteamine (amino-thiol), triethylsilane (reactive hydrosilane), hexamethyldisilazane, nonamethyltrisilazane, octamethylcyclotetrasilazane (reactive amines). The results showed that the reactive amines were good additive ligands for enhancing the fluorescence of CdSe/ZnS quantum dots dispersed in the silicone resins, providing 1.2-2.48 Im/W and 4.2-5.56% higher luminous efficiency and photoluminescence conversion efficiency, respectively. We speculate that these reactive amines donate electrons to the surface electron traps, thereby reducing charge recombination. In addition, quantum dots aggregate to form quantum dot clusters with a relatively homogeneously dispersed in the silicone resin matrices, showing good emission properties due to surface passivation and good colloidal stability with the addition of silazane compounds to the resin. Furthermore, the addition of silazane compounds to quantum dots-silicone resin system also shows the improved thermal stability of the as-synthesized nanocomposites.
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Sigma-Aldrich
Zinc sulfide, pieces, 3-12 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc sulfide, purum, 97% (from Zn)
Sigma-Aldrich
Zinc sulfide, powder, 10 μm, 99.99% trace metals basis