MilliporeSigma
  • Lysine 313 of T-box is crucial for modulation of protein stability, DNA binding, and threonine phosphorylation of T-bet.
Lysine 313 of T-box is crucial for modulation of protein stability, DNA binding, and threonine phosphorylation of T-bet.
Journal of immunology (Baltimore, Md. : 1950) (2013-04-26)
Eun Jung Jang, Hye Ryeon Park, Jeong-Ho Hong, Eun Sook Hwang
ABSTRACT
A T-box-containing protein expressed in T cells (T-bet) is a key transcription factor involved in the regulation of Th cell differentiation. Although T-bet-deficient CD4(+) T cells fail to produce IFN-γ and typically differentiate into Th2 cells in vitro, ectopic overexpression of T-bet elevates IFN-γ and suppresses production of IL-2 and Th2 cytokines through different mechanisms. Despite the importance of the T-bet protein level, the regulatory mechanisms that control T-bet protein stability are largely unknown. In this study, we found that T-bet underwent proteasomal degradation via ubiquitination at Lys-313. Despite its robust accumulation following lysine mutation, T-bet(K313R) failed to increase IFN-γ production because of diminished DNA binding activity, as demonstrated in the crystal structure of T-bet-DNA complex. Strikingly, T-bet(K313R) entirely lost the ability to suppress IL-2 production and Th2 cell development; this was due to loss of its interaction with NFAT1. We further identified that the T-bet(K313R) reduced the phosphorylation of T-bet at Thr-302, and that threonine phosphorylation was essential for T-bet interaction with NFAT1 and suppression of NFAT1 activity. Retroviral transduction of T-bet(T302A) into T-bet-deficient cells restored IFN-γ levels compared with those induced by wild-type T-bet, but this mutant failed to inhibit IL-2 and Th2 cytokine production. Collectively, these data show that Lys-313 in the T-box domain is essential for controlling T-bet protein stability via ubiquitin-dependent degradation, T-bet binding to the IFN-γ promoter, and for the interaction with and suppression of NFAT1. Thus, multiple posttranslational modifications of T-bet are involved in fine-tuning cytokine production during Th cell development.
Product Description
SAFC
L-Threonine
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material