Merck
  • [Ir(P-OP)]-catalyzed asymmetric hydrogenation of diversely substituted C═N-containing heterocycles.
[Ir(P-OP)]-catalyzed asymmetric hydrogenation of diversely substituted C═N-containing heterocycles.
Organic letters (2013-04-12)
José Luis Núñez-Rico, Anton Vidal-Ferran
ABSTRACT
Iridium(I) complexes of enantiomerically pure phosphine-phosphite ligands ([Ir(Cl)(cod)(P-OP)]) efficiently catalyze the enantioselective hydrogenation of diverse C═N-containing heterocyclic compounds (benzoxazines, benzoxazinones, benzothiazinones, and quinoxalinones; 25 examples, up to 99% ee). A substrate-to-catalyst ratio as high as 2000:1 was reached.
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis