Merck
  • Prophylactic salpingectomy in premenopausal low-risk women for ovarian cancer: primum non nocere.
Prophylactic salpingectomy in premenopausal low-risk women for ovarian cancer: primum non nocere.
Gynecologic oncology (2013-04-06)
Michele Morelli, Roberta Venturella, Rita Mocciaro, Annalisa Di Cello, Erika Rania, Daniela Lico, Pietro D'Alessandro, Fulvio Zullo
ABSTRACT
The objective of this study is to compare ovarian function and surgical outcomes between patients affected by benign uterine pathologies submitted to total laparoscopic hysterectomy (TLH) plus salpingectomy and women in which standard TLH with adnexal preservation was performed. We retrospectively compared data of 79 patients who underwent TLH plus bilateral salpingectomy (group A), with those of 79 women treated by standard TLH without adnexectomy (sTLH) (group B). Ovarian reserve modification, expressed as the difference between 3 months post-operative and pre-operative values of Anti-Müllerian Hormone (AMH), Follicle Stimulating Hormone (FSH), Antral Follicle Count (AFC), mean ovarian diameters and Peak Systolic Velocity (PSV), was recorded for each patient. For each surgical procedure, operative time, variation of hemoglobin level (ΔHb), postoperative hospital stay, postoperative return to normal activity, and complication rate were recorded as secondary outcomes. According to our post-hoc analysis, this equivalence study resulted to have a statistical power of 96.8%. Significant difference was not observed between groups with respect to ΔAMH (p=0.35), ΔFSH (p=0.15), ΔAFC (p=0.09), Δ mean ovarian diameters (p=0.57) and ΔPSV (p=0.61). In addition, secondary outcomes such as operative time (p=0.79), ΔHb (p=0.41), postoperative hospital stay (p=0.16), postoperative return to normal activity (p=0.11) and complication rate also did not show any significant difference. The addition of bilateral salpingectomy to TLH for prevention of ovarian cancer in women who do not carry a BRCA1/2 mutations do not show negative effects on the ovarian function. In addition, no perioperative complications are related to the salpingectomy step in TLH.
Sigma-Aldrich
Follicle Stimulating Hormone from human pituitary, ~7,000 IU/mg (powder)