Merck
  • Effect of human urine on the tensile strength of sutures used for hypospadias surgery.
Effect of human urine on the tensile strength of sutures used for hypospadias surgery.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS (2013-04-06)
Ryan L Kerstein, Tina Sedaghati, Alexander M Seifalian, Norbert Kang
ABSTRACT
Hypospadias is the most common congenital condition affecting between 1 in 250 and 300 live births. Even in experienced hands, surgery to repair this congenital defect can have a high complication rate. Wound dehiscence is reported to occur in 5% and fistula formation in 6%-40% depending on technique. The choice of suture material has been shown to affect the complication rate although there is (currently) no consensus about the best suture material to use. Ideally, the sutures used for urethroplasty should be absorbable while maintaining sufficient mechanical strength to support the wounds until they are self-supporting and able to resist urinary flow. Previous studies have compared the effects of human urine on different suture materials especially catgut. However, catgut is now banned in Europe. Our study examined the tensile and breaking strength as well as rate of degradation for four types of absorbable suture now commonly used for hypospadias repairs in the UK. We examined the effect of prolonged storage (up to 27 days) in human urine on 6/0 gauge Vicryl, Vicryl Rapide, Monocryl and polydioxanone (PDS) sutures. These four suture materials are commonly used by the senior plastic consultant surgeon (NK) for hypospadias repairs. 50 mm sections of these suture materials were stored in either urine or saline as control. At specified time points, each suture was placed in a uniaxial load testing machine to assess the stress-strain profile and the mechanical load required to break the suture was measured. Exposure to urine reduced the tensile and breaking strength of all the suture materials tested. PDS demonstrated the greatest resilience. Vicryl Rapide was the weakest suture and degraded completely by day 6. Vicryl and Monocryl had similar degradation profiles, but Vicryl retained more of its tensile strength for longer. There is a balance to be struck between the duration that a suture material must remain in any surgical wound and the risk that it causes foreign body effects. The results of this study suggest that Vicryl has the best characteristics for urethroplasty of the four suture materials tested.
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000