Merck
  • In vitro stability of free and glucuronidated cannabinoids in blood and plasma following controlled smoked cannabis.
In vitro stability of free and glucuronidated cannabinoids in blood and plasma following controlled smoked cannabis.
Clinical chemistry (2013-03-23)
Karl B Scheidweiler, David M Schwope, Erin L Karschner, Nathalie A Desrosiers, David A Gorelick, Marilyn A Huestis
ABSTRACT
Blood and plasma cannabinoid stability is important for test interpretation and is best studied in authentic rather than fortified samples. Low and high blood and plasma pools were created for each of 10 participants after they smoked a cannabis cigarette. The stabilities of Δ(9)-tetrahydrocannabinol (THC), 11-hydroxy-THC (11-OH-THC), 11-nor-9-carboxy-THC (THCCOOH), cannabidiol (CBD), cannabinol (CBN), THC-glucuronide, and THCCOOH-glucuronide were determined after 1 week at room temperature; 1, 2, 4, 12, and 26 (±2) weeks at 4 °C; and 1, 2, 4, 12, 26 (±2), and 52 (±4) weeks at -20 °C. Stability was assessed by Friedman test. Numbers of THC-glucuronide and CBD-positive blood samples were insufficient to assess stability. In blood, 11-OH-THC and CBN were stable for 1 week at room temperature, whereas THC and THCCOOH-glucuronide decreased and THCCOOH increased. In blood, THC, THCCOOH-glucuronide, THCCOOH, 11-OH-THC, and CBN were stable for 12, 4, 4, 12, and 26 weeks, respectively, at 4 °C and 12, 12, 26, 26, and 52 weeks at -20 °C. In plasma, THC-glucuronide, THC, CBN, and CBD were stable for 1 week at room temperature, whereas THCCOOH-glucuronide and 11-OH-THC decreased and THCCOOH increased. In plasma, THC-glucuronide, THC, THCCOOH-glucuronide, THCCOOH, 11-OH-THC, CBN, and CBD were stable for 26, 26, 2, 2, 26, 12, and 26 weeks, respectively, at 4 °C and 52, 52, 26, 26, 52, 52, and 52 weeks, respectively, at -20 °C. Blood and plasma samples should be stored at -20 °C for no more than 3 and 6 months, respectively, to assure accurate cannabinoid quantitative results.
Product Description
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-trans-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cannabidiol solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Supelco
Cannabidiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cannabinol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cannabidiol solution, ~10 mg/mL in ethanol, analytical standard, for drug analysis