Merck
  • Peptide directed transmembrane transport and nuclear localization of Ru(II) polypyridyl complexes in mammalian cells.
Peptide directed transmembrane transport and nuclear localization of Ru(II) polypyridyl complexes in mammalian cells.
Chemical communications (Cambridge, England) (2013-02-26)
Lorraine Blackmore, Roisin Moriarty, Ciaran Dolan, Kellie Adamson, Robert J Forster, Marc Devocelle, Tia E Keyes
ABSTRACT
The targeted delivery of luminescent Ru(II) polypyridyl complexes to the nucleus of live mammalian cells by a Nuclear Localisation Signal (NLS) peptide; NF-κB is demonstrated.
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis