Merck
  • CdTe nanobioprobe based optoelectrochemical immunodetection of diabetic marker HbA1c.
CdTe nanobioprobe based optoelectrochemical immunodetection of diabetic marker HbA1c.
Biosensors & bioelectronics (2013-02-19)
Adity Chopra, Satish Tuteja, Naresh Sachdeva, K K Bhasin, Vijayender Bhalla, C Raman Suri
ABSTRACT
Highly luminescent water soluble CdTe quantum dots (QDs) were synthesized and conjugated with anti-HbA1c antibody to generate specific nanobioprobe. A sandwich immunoassay model was employed using capture HbA1c antibody as a specific receptor molecule and the QD-labeled secondary antibody as a dual (fluorescence cum electrochemical) tracer to quantify the concentration of HbA1c. A linear increase in current was observed for HbA1c analytical standards with a R(2) value of 0.990 and coefficient of variance ~5%. A comparison between HPLC and dual immunoassay for clinical samples showed a correlation coefficient of 89% and 96% for fluorescence and electrochemical detection methods respectively. The QD-based immunoassay shows great promise for rapid reproducible and cost effective analysis of HbA1c in clinical samples.
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, pieces, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis