MilliporeSigma
  • Struma ovarii simulating ovarian sertoli cell tumor: a case report with literature review.
Struma ovarii simulating ovarian sertoli cell tumor: a case report with literature review.
International journal of clinical and experimental pathology (2013-02-16)
Yan Ning, Fanbin Kong, Janiel M Cragun, Wenxin Zheng
ABSTRACT
Struma ovarii, as a monodermal variant of ovarian teratoma, constitutes about less than 3% of ovarian teratomas. It is difficult to be macroscopically recognized. Multiple appearances under microscope serve as another reason to mislead the accurate pathologic evaluation. Here, we report an unusual case of struma ovarii occurred in a 77 years old woman, which is currently known as the oldest age for this disease. The frozen section morphologically showed sex cord like elements and was suspicious for a sex-cord stromal tumor, probably a Sertoli cell tumor. Final pathological diagnosis was confirmed as struma ovarii based on the typical morphologic thyroid follicles and immunohistochemical staining results.
Sigma-Aldrich
Thyroglobulin human
Pricing and availability is not currently available.
Sigma-Aldrich
Thyroglobulin from porcine thyroid gland, powder
Pricing and availability is not currently available.