Merck
  • Synthesis and swelling properties of β-cyclodextrin-based superabsorbent resin with network structure.
Synthesis and swelling properties of β-cyclodextrin-based superabsorbent resin with network structure.
Carbohydrate polymers (2013-02-13)
Zhanhua Huang, Shouxin Liu, Guizhen Fang, Bin Zhang
ABSTRACT
A biodegradable, β-cyclodextrin-based superabsorbent resin was synthesized by the inverse suspension method. The microstructure, chemical structure, and thermal performance of the resin were characterized by scanning electron microscopy, Fourier transform-infrared spectroscopy, and differential scanning calorimetry. The effects of the synthesis conditions (dosage of cross-linking agent, mass ratios of acrylic acid to acrylamide, mass ratios of β-cyclodextrin to total monomer, neutralization degree, initiator dosage, and reaction time) were optimized to achieve a resin with a maximum swelling capacity. The water absorbency of the optimized resin in distilled water was 1544.76 g/g and that in 0.9 wt.% NaCl was 144.52 g/g. The resin, which is thermoplastic as well as pH-sensitive, had good salt resistance and underwent a maximum in swelling with time in CaCl(2) and AlCl(3) solutions. The fracture surface of the dry resin contained many pores. After swelling, the internal hydrogel showed a typical three-dimensional network structure. The biodegradation of the resin reached 71.2% after 18 days treatment at 30 °C with Lentinus edodes.
Sigma-Aldrich
Chlorine, ≥99.5%
Sigma-Aldrich
Chlorine, puriss., ≥99.8%