Merck
  • The reaction of ethyl 2-oxo-2H-chromene-3-carboxylate with hydrazine hydrate.
The reaction of ethyl 2-oxo-2H-chromene-3-carboxylate with hydrazine hydrate.
Molecules (Basel, Switzerland) (2013-02-08)
Hatem A Abdel-Aziz, Tilal Elsaman, Mohamed I Attia, Amer M Alanazi
ABSTRACT
Although salicylaldehyde azine (3) was reported in 1985 as the single product of the reaction of ethyl 2-oxo-2H-chromene-3-carboxylate (1) with hydrazine hydrate, we identified another main reaction product, besides 3, which was identified as malono-hydrazide (4). In the last two decades, however, some articles have claimed that this reaction afforded exclusively hydrazide 2 and they have reported the use of this hydrazide 2 as a precursor in the syntheses of several heterocyclic compounds and hydrazones 6. We reported herein a study of the formation of 2 and a facile route for the synthesis of the target compounds N'-arylidene-2-oxo-2H-chromene-3-carbohydrazides 6a-f.
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%