Merck
  • Phenothiazine as an aromatic capping group to construct a short peptide-based 'super gelator'.
Phenothiazine as an aromatic capping group to construct a short peptide-based 'super gelator'.
Chemical communications (Cambridge, England) (2013-01-31)
Caiwen Ou, Jianwu Zhang, Xiaoli Zhang, Zhimou Yang, Minsheng Chen
ABSTRACT
We report a 'super-gelator' of a short peptide capped with phenothiazine acetic acid.
Sigma-Aldrich
Phenothiazine, ≥98%
Sigma-Aldrich
Phenothiazine, purum, ≥98.0% (GC)