Merck
  • Dendrimers as Nd3+ ligands: effect of generation on the efficiency of the sensitized lanthanide emission.
Dendrimers as Nd3+ ligands: effect of generation on the efficiency of the sensitized lanthanide emission.
Chemistry, an Asian journal (2013-01-25)
Zeena S Pillai, Paola Ceroni, Manja Kubeil, Jan-Martin Heldt, Holger Stephan, Giacomo Bergamini
ABSTRACT
We have designed two novel dendrimers with cyclam cores with appended poly(amido amine) (PAMAM) dendrons, decorated at the periphery with four and eight dansyl chromophores, respectively. The photophysical properties of the dendrimers and their Nd(3+) complexes have been investigated. The energy-transfer efficiency to the lanthanide ions from these dendrimers has been studied as a function of the generation. It has been observed that an increase in the dendrimer generation as well as the number of amide units enhances the energy transfer to the lanthanide ion.
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)