Merck
  • Two-dimensional magnetic clusters in La(n)(Ti(1-x)Fe(x))(n)O(3n+2) (n = 5 with x = 0.2 and n = 6 with x = 0.33).
Two-dimensional magnetic clusters in La(n)(Ti(1-x)Fe(x))(n)O(3n+2) (n = 5 with x = 0.2 and n = 6 with x = 0.33).
Journal of physics. Condensed matter : an Institute of Physics journal (2013-01-19)
Alexander Wölfel, Frank Lichtenberg, Sander van Smaalen
ABSTRACT
Crystal structure analyses of the layered compounds La(n)(Ti(1-x)Fe(x))(n)O(3n+2), with nominal compositions x = 0.2 for n = 5 and x = 0.33 for n = 6, show that the iron is concentrated at the centers of the slabs. The spatial arrangements of the iron ions can be approximated by two-dimensional square lattices with strong magnetic interactions between neighboring sites, albeit with fractional occupancies of the sites of an average of 0.67 magnetic ions in n = 6 and 0.42 magnetic ions in n = 5 compounds. A previously described (Lichtenberg et al 2008 Prog. Solid State Chem. 36 253) crossover of magnetic behavior of n = 6 at room temperature is explained by the formation of two-dimensional, ferromagnetically organized magnetic clusters with an average size of 52 Fe(3+) ions. The absence of long-range magnetic order follows from the fractional occupancy of the sites by the magnetic ions. The lower concentration of magnetic ions in n = 5 explains why crossover behavior is not found and clusters do not form in that compound.
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis