Merck
Key structure of brij for overcoming multidrug resistance in cancer.
Biomacromolecules (2013-01-15)
Jingling Tang, Yongjun Wang, Dun Wang, Yuhua Wang, Zhenghong Xu, Kelly Racette, Feng Liu
ABSTRACT
Multidrug resistance (MDR) is a major barrier to the chemotherapy treatment of many cancers. However, some nonionic surfactants, for example, Brij, have been shown to restore the sensitivity of MDR cells to such drugs. The aim of this study was to explore the reversal effect of Brij on MDR tumor cells and elucidate its potential mechanism. Our data indicate that the structure of Brij surfactants plays an important role in overcoming MDR in cancer, that is, modified hydrophilic-lipophilic balance (MHLB, the ratio of the number (n) of hydrophilic repeating units of ethylene oxide (EO) to the number (m) of carbons in the hydrophobic tail (CH(2))). Cell viability of cells treated with paclitaxel (PTX) nanocrystals (NCs) formulated with Brij showed positive correlations with MHLB (R(2) = 0.8195); the higher the ratio of Brij to PTX in NCs, the higher cytotoxicity induced by the PTX NCs. Significant increases in intracellular accumulation of (3)H-PTX (P-gp substrate) were observed in an MDR cell line (H460/taxR cells) treated with Brij 78 (MHLB = 1.11) and Brij 97 (MHLB = 0.6). After treatments with Brij 78 and Brij 97, the levels of intracellular ATP were decreased and verapamil-induced ATPase activities of P-gp were inhibited in multidrug resistant cells. The responses of the cells to Brij 78 and Brij 97 in ATP depletion studies correlated with the cell viability induced by PTX/Brij NCs and intracellular accumulation of (3)H-PTX. Brij 78 and Brij 97 could not alter the levels of P-gp expression detected by Western blotting. These findings may provide some insight into the likelihood of further development of more potent P-gp inhibitors for the treatment of MDR in cancer.
Sigma-Aldrich
Brij® S10, average Mn ~711
Sigma-Aldrich
Brij® 93, average Mn ~357
Sigma-Aldrich
Brij® O10
Sigma-Aldrich
Brij® S20
Sigma-Aldrich
ECO Brij® S20
Sigma-Aldrich
ECO Brij® S100, average Mn ~4,670
Sigma-Aldrich
ECO Brij® S10, average Mn ~711
Sigma-Aldrich
ECO BRIJ® O20, average Mn ~1,150
Sigma-Aldrich
ECO BRIJ® O10
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
BRIJ® O20, average Mn ~1,150
Sigma-Aldrich
Brij® S 100, average Mn ~4,670
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder