Merck
  • Effect of calcination temperature on the activity of solid Ca/Al composite oxide-based alkaline catalyst for biodiesel production.
Effect of calcination temperature on the activity of solid Ca/Al composite oxide-based alkaline catalyst for biodiesel production.
Bioresource technology (2012-12-01)
Yong-Lu Meng, Bo-Yang Wang, Shu-Fen Li, Song-Jiang Tian, Min-Hua Zhang
ABSTRACT
A solid Ca/Al composite oxide-based alkaline catalyst containing Ca(12)Al(14)O(33) and CaO was prepared by chemical synthesis and thermal activation from sodium aluminate solution and calcium hydroxide emulsion. The effect of calcination temperatures ranging from 120 °C to 1000 °C on activity of the catalyst was investigated. The catalyst calcined at 600 °C showed the highest activity with >94% yield of fatty acid methyl esters (i.e. biodiesel) when applied to the transesterification of rapeseed oil at a methanol:oil molar ratio of 15:1 at 65 °C for 3h. Structure and properties of the catalyst were studied and the characterizations with XRD, TGA, FTIR, BET, and SEM demonstrated that the performance of the catalyst was closely related to its specific surface area and crystalline structure. In particular, the generation of crystalline Ca(12)Al(14)O(33) improved the catalytic activity due its synergistic effect with CaO.
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, reagent grade
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%