Merck
Which polyesters can mimic polyethylene?
Macromolecular rapid communications (2012-11-20)
Florian Stempfle, Patrick Ortmann, Stefan Mecking
ABSTRACT
Self-metathesis of erucic acid by [(PCy(3))(η-C-C(3)H(4)N(2)Mes(2))Cl(2)Ru = CHPh] (Grubbs second- generation catalyst) followed by catalytic hydrogenation and purification via the ester yields 1,26-hexacosanedioate (>99% purity). Polyesterification with 1,26-hexacosanediol, generated from the diester, affords polyester-26,26, which features a T(m) of 114 °C (T(c) = 92 °C, ΔH(m) = 160 J g(-1)). Ultralong-chain model polyesters-38,23 (T(m) = 109 °C) and -44,23 (T(m) = 111 °C), generated via multistep procedures including acyclic diene metathesis polymerization, underline that melting points of such aliphatic polyesters do not gradually increase with methylene sequence chain length. Available data suggest that to mimic linear polyethylenes thermal properties, even longer sequences, amounting to at least four times a fatty acid chain, fully incorporated in a linear fashion are required.
Product Description
Supelco
Erucic acid, analytical standard
Sigma-Aldrich
Erucic acid, ≥99% (capillary GC)
Sigma-Aldrich
Erucic acid, technical, ~90% (GC)