Merck
  • Anti-inflammatory effects of egg white combined with chalcanthite in lipopolysaccharide-stimulated BV2 microglia through the inhibition of NF-κB, MAPK and PI3K/Akt signaling pathways.
Anti-inflammatory effects of egg white combined with chalcanthite in lipopolysaccharide-stimulated BV2 microglia through the inhibition of NF-κB, MAPK and PI3K/Akt signaling pathways.
International journal of molecular medicine (2012-11-07)
Eun A Choi, Hye Young Park, Hwa-Seung Yoo, Yung Hyun Choi
ABSTRACT
Egg white-chalcanthite (EWCC) is a mixture of egg white and chalcanthite prepared by roasting chalcanthite (which is a natural mineral mainly composed of CuSO4•5H2O) to the point of dehydration, pulverizing the dehydrated chalcanthite and then mixing the pulverized chalcanthite to react with egg white to trigger a reaction. When egg white-chalcanthite is prepared in this manner, the toxicity of chalcanthite is neutralized by the egg white, so that the toxicity is reduced or removed and the pharmaceutical properties are increased. However, the cellular and molecular mechanisms underlying the pharmacological activity of EWCC remain poorly understood. In this study, we investigated the inhibitory effects of EWCC on the production of lipopolysaccharide (LPS)-induced pro-inflammatory mediators in BV2 microglia. Our data indicated that the EWCC treatment significantly inhibited the excessive production of nitric oxide and prostaglandin E2 in LPS-stimulated BV2 microglia in a concentration-dependent manner without causing cytotoxicity. It also attenuated the expression of inducible nitric oxide synthase, cyclooxygenase-2 and pro-inflammatory cytokines, including interleukin-1β and tumor necrosis factor-α. Moreover, EWCC exhibited anti-inflammatory properties by the suppression of nuclear factor‑κB (NF-κB) activation by blocking IκB-α degradation, downregulation of extracellular signal-regulated kinase, c-Jun N-terminal kinase, p38 mitogen-activated protein kinase (MAPK) and phosphoinositide 3-kinase (PI3K)/Akt pathways. Our results indicate that the inhibitory effects of EWCC on LPS-stimulated inflammatory mediator production in BV2 microglia are associated with the suppression of the NF-κB, MAPK and PI3K/Akt signaling pathways. These findings suggest that EWCC may offer a substantial therapeutic potential for the treatment of neurodegenerative diseases that are accompanied by microglial activation.
Sigma-Aldrich
Copper(II) sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99-100.5% (based on anhydrous substance)
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Sigma-Aldrich
Copper(II) sulfate, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis