Merck
  • Radical amination of C(sp3)-H bonds using N-hydroxyphthalimide and dialkyl azodicarboxylate.
Radical amination of C(sp3)-H bonds using N-hydroxyphthalimide and dialkyl azodicarboxylate.
The Journal of organic chemistry (2012-11-02)
Yuuki Amaoka, Shin Kamijo, Tamaki Hoshikawa, Masayuki Inoue
ABSTRACT
A direct conversion of C(sp(3))-H bonds to C(sp(3))-N bonds has been achieved by utilizing catalytic N-hydroxyphthalimide (NHPI) and stoichiometric dialkyl azodicarboxylate. NHPI functions as a precursor of the electron-deficient phthalimide N-oxyl radical (PINO) to abstract hydrogens, and dialkyl azodicarboxylate acts as a trapping agent of the resultant carbon radical to generate the hydrazine derivatives. This C-H amination proceeds in a highly chemoselective manner with a wide applicability to functionalize benzylic, propargylic, and aliphatic C-H bonds. Furthermore, the obtained hydrazine compounds were readily converted to the corresponding carbamates or amines. Hence, the present protocol for direct introduction of the nitrogen functionality serves as a powerful tool for efficient construction of nitrogen-substituted natural products and pharmaceuticals.
Sigma-Aldrich
N-Hydroxyphthalimide, 97%
Pricing and availability is not currently available.