Merck
  • Immobilization of bacterial luciferase into poly(N-isopropylacrylamide) film for electrochemical control of a bioluminescence reaction.
Immobilization of bacterial luciferase into poly(N-isopropylacrylamide) film for electrochemical control of a bioluminescence reaction.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2012-10-13)
Yoji Kawanami, Shinya Yamasaki, Shuto Yamada, Kô Takehara
ABSTRACT
A poly(N-isopropylacrylamide) film was modified on an indium-tin oxide electrode in order to immobilize bacterial luciferase (BL) on the electrode surface. By using the modified electrode, flavin mononucleotide (FMN) was electrochemically reduced to FMNH(2), which is one of the substrates of the BL luminescence reaction, to control the bioluminescence reaction by BL. The BL reaction in the modified film could be promoted and controlled by the electrochemical generation of FMNH(2). This BL luminescence system was evaluated as a model system for the inhibitory assay of hydrophobic molecules on protein functions.
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(N-isopropylacrylamide), carboxylic acid terminated, average Mn 10,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
1-Dodecanol, Selectophore, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(N-isopropyl acrylamide), NHS ester end functionalized, average Mn 5,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%