Merck
  • Selective sensing of mercury(II) using PVC-based membranes incorporating recently synthesized 1,3-alternate thiacalix[4]crown ionophore.
Selective sensing of mercury(II) using PVC-based membranes incorporating recently synthesized 1,3-alternate thiacalix[4]crown ionophore.
Environmental science and pollution research international (2012-10-12)
Rakesh Kumar Mahajan, Ajar Kamal, Naresh Kumar, Vandana Bhalla, Manoj Kumar
ABSTRACT
The construction and electrodes characteristics of poly(vinylchloride) (PVC)-based polymeric membrane electrode (PME) and coated graphite electrode (CGE), incorporating 1,3-alternate thiacalix[4]crown as ionophore for estimation of Hg(II) ions, are reported here. The best potential response was observed for PME-1 having membrane composition of: ionophore (6.2 mg), PVC (100.0 mg), 2-nitrophenyl octyl ether (2-NPOE; 200.0 mg), and sodium tetraphenyl borate (NaTPB; 2.0 mg); for CGE-1 with the membrane composition: ionophore (3.5 mg), PVC (40.0 mg), 2-NPOE (80.0 mg), and NaTPB (2.0 mg). The electrodes exhibits Nernstian slope of 29.16 mV/decade with PME-1 and 30.39 mV/decade with CGE-1 for Hg(II) ions over wide concentration range, i.e., 1.0 × 10(-1) to 5.0 × 10(-6) M with PME-1 and 1.0 × 10(-1) to 5.0 × 10(-7) M with CGE-1. Lower detection limits were found to be 9.77 × 10(-6) M for PME-1 and 7.76 × 10(-7) M for CGE-1 with response time varying from 10 to 20 s. Also, these electrodes work within pH range of 2.0-6.0 for PME-1 and 1.5-6.5 for CGE-1. Overall, CGE-1 has been found to be better than PME-1. CGE-1 has been used as indicator electrode for the potentiometric titration of Hg(II) ions with EDTA as well as successfully applied for determination of Hg(II) content in wastewater, insecticide, dental amalgam, and ayurvedic medicines samples with very good performance (0.9974 correlation coefficient in the comparison against volumetric method).
Product Description
Supelco
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight
Sigma-Aldrich
Poly(vinyl chloride), low molecular weight
Supelco
Poly(vinyl chloride), Selectophore, high molecular weight
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~43,000, average Mn ~22,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard